siRNA Details
Gene Symbol: Sidt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289668.1 | NP_001276597.1 | 22, 23 | 2473 | ||
NM_172257.4 | NP_758461.1 | 21, 22 | 2410 | |||
GenBank mRNA | AK155875.1 | - | 21, 22 | 2314 | ||
BC006873.1 | - | 8, 9 | 747 | |||
BC023957.1 | - | 21, 22 | 2414 | 5321190 | ||
BC051101.1 | - | 22, 23 | 2201 | 5052849 | ||
BQ749207.1 | - | 4, 5 | 391 | |||
CA315096.1 | - | 9, 10 | 809 |
