siRNA Details
| Gene Symbol: Slc25a29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181328.3 | NP_851845.1 | 4 | 394 | ||
| GenBank mRNA | AK047628.1 | - | 4 | 395 | ||
| AK087057.1 | - | 4 | 381 | |||
| AK148350.1 | - | 4 | 372 | |||
| AK160906.1 | - | 4 | 343 | |||
| AK180537.1 | - | 2 | 122 | |||
| AK181345.1 | - | 2 | 122 | |||
| BC006711.1 | - | 4 | 267 | 3584570 | ||

Slc25a29
Genomic Map