siRNA Details
Gene Symbol: Csnk1g1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173185.2 | NP_775277.2 | 4 | 625 | ||
GenBank mRNA | AK050473.1 | - | 4 | 606 | ||
AK090262.1 | - | 4 | 626 | |||
AK149310.1 | - | 5 | 643 | |||
BC064645.1 | - | 3 | 641 | 5363779 |
