siRNA Details
Gene Symbol: Mob3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178061.5 | NP_835162.1 | 2 | 609 | ||
GenBank mRNA | AK039841.1 | - | 2 | 455 | ||
AK087285.1 | - | 2 | 610 | |||
AK155217.1 | - | 2 | 610 | |||
AK170973.1 | - | 2 | 603 | |||
BC020028.1 | - | 2 | 591 | 4007373 |
