siRNA Details
| Gene Symbol: Chtf8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145412.3 | NP_663387.3 | 3 | 214 | ||
| GenBank mRNA | AK030924.1 | - | 3 | 213 | ||
| AK088257.1 | - | 3 | 223 | |||
| AK165622.1 | - | 3 | 196 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Chtf8| Gene Symbol: Chtf8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145412.3 | NP_663387.3 | 3 | 214 | ||
| GenBank mRNA | AK030924.1 | - | 3 | 213 | ||
| AK088257.1 | - | 3 | 223 | |||
| AK165622.1 | - | 3 | 196 | |||