siRNA Details
| Gene Symbol: Hip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146001.2 | NP_666113.2 | 20, 21 | 2097 | ||
| GenBank mRNA | AK147246.1 | - | 20, 21 | 2052 | ||
| AK147269.1 | - | 20, 21 | 2052 | |||
| AK220182.1 | - | 7, 8 | 798 | |||
| BC017516.1 | - | 20, 21 | 2090 | 4504441 | ||

Hip1
Genomic Map