siRNA Details
| Gene Symbol: Wipf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289722.1 | NP_001276651.1 | 3 | 292 | ||
| NM_001289723.1 | NP_001276652.1 | 3 | 257 | |||
| NM_153138.4 | NP_694778.1 | 3 | 287 | |||
| GenBank mRNA | AJ437262.1 | - | 2 | 88 | ||
| AK031046.1 | - | 3 | 254 | |||
| AK156547.1 | - | 3 | 292 | |||
| BC049788.1 | - | 3 | 210 | 6514406 | ||
| BY200699.1 | - | 3 | 258 | |||
| BY252790.1 | - | 3 | 288 | |||
| BY285630.1 | - | 3 | 247 | |||

Wipf1
Genomic Map