siRNA Details
| Gene Symbol: Brinp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145807.1 | NP_001139279.1 | 8 | 1755 | ||
| NM_153539.3 | NP_705767.3 | 8 | 2113 | |||
| GenBank mRNA | AK046878.1 | - | 8 | 2114 | ||
| AK083233.1 | - | 9 | 1804 | |||
| BC030498.1 | - | 7 | 1731 | 5400263 | ||
| BC099956.1 | - | 8 | 2090 | 30101722 | ||

Brinp3
Genomic Map