siRNA Details
| Gene Symbol: Nav1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173437.2 | NP_775613.2 | 16 | 3574 | ||
| GenBank mRNA | AF307453.1 | - | 16 | 3934 | ||
| AK053255.1 | - | 14 | 2553 | |||
| AK082667.1 | - | 14 | 2487 | |||
| AK122458.1 | - | 17 | 3641 | |||
| BC059840.1 | - | 16 | 3717 | 6836774 | ||
| CB523329.1 | - | 1 | 24 | |||
| CN455316.1 | - | 7 | 559 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK147844.1 | - | 3290 | Chr. - - [-] on Build | ||

Nav1
Genomic Map