siRNA Details
| Gene Symbol: Nhsl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163592.1 | NP_001157064.1 | 6 | 3485 | ||
| NM_173390.3 | NP_775566.3 | 5 | 4008 | |||
| GenBank mRNA | AK220386.1 | - | 1 | 53 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nhsl1| Gene Symbol: Nhsl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163592.1 | NP_001157064.1 | 6 | 3485 | ||
| NM_173390.3 | NP_775566.3 | 5 | 4008 | |||
| GenBank mRNA | AK220386.1 | - | 1 | 53 | ||