siRNA Details
| Gene Symbol: Nhsl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173390.3 | NP_775566.3 | 1 | 707 | ||
| GenBank mRNA | AK017530.1 | - | 1 | 708 | ||
| AK038892.1 | - | 1 | 683 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nhsl1| Gene Symbol: Nhsl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173390.3 | NP_775566.3 | 1 | 707 | ||
| GenBank mRNA | AK017530.1 | - | 1 | 708 | ||
| AK038892.1 | - | 1 | 683 | |||