siRNA Details
Gene Symbol: Stox1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033260.1 | NP_001028432.1 | 3 | 1125 | ||
GenBank mRNA | AK028920.1 | - | 3 | 891 | ||
BC137978.1 | - | 3 | 1125 | 8860997 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Stox1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033260.1 | NP_001028432.1 | 3 | 1125 | ||
GenBank mRNA | AK028920.1 | - | 3 | 891 | ||
BC137978.1 | - | 3 | 1125 | 8860997 |