siRNA Details
Gene Symbol: Ube2d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145420.2 | NP_663395.1 | 1, 2 | 153 | ||
GenBank mRNA | AK043167.1 | - | 1, 2 | 109 | ||
AK144468.1 | - | 1, 2 | 108 | |||
AK148364.1 | - | 1, 2 | 141 | |||
BC019464.1 | - | 1, 2 | 124 | 4205941 |
