siRNA Details
| Gene Symbol: Ilvbl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173751.4 | NP_776112.1 | 11, 12 | 1487 | ||
| GenBank mRNA | AK087956.1 | - | 10, 11 | 1361 | ||
| AK146075.1 | - | 10, 11 | 1415 | |||
| AK169899.1 | - | 10, 11 | 1413 | |||
| AK218076.1 | - | 1 | 37 | |||
| BC052054.1 | - | 10, 11 | 1821 | |||
| BC056459.1 | - | 11, 12 | 1445 | 5718157 | ||

Ilvbl
Genomic Map