siRNA Details
| Gene Symbol: Wdr18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175450.4 | NP_780659.2 | 2 | 284 | ||
| GenBank mRNA | AK032251.1 | - | 2 | 282 | ||
| AK050106.1 | - | 2 | 282 | |||
| AK082695.1 | - | 2 | 286 | |||
| AK087505.1 | - | 1 | 915 | |||
| AK133716.1 | - | 2 | 285 | |||
| AK160713.1 | - | 2 | 282 | |||
| AK163465.1 | - | 1 | 912 | |||
| AK166113.1 | - | 282 | ||||
| AK168691.1 | - | 2 | 282 | |||
| BC095984.1 | - | 2 | 284 | 4978914 | ||

Wdr18
Genomic Map