siRNA Details
| Gene Symbol: BC089597 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145424.2 | NP_663399.2 | 1 | 146 | ||
| GenBank mRNA | AK144071.1 | - | 1 | 135 | ||
| AY053571.1 | - | 1 | 66 | |||
| AY053572.1 | - | 1 | 146 | |||
| BC018263.1 | - | 1 | 142 | 4239862 | ||
| BC089597.1 | - | 1 | 120 | 30120654 | ||

BC089597
Genomic Map