siRNA Details
Gene Symbol: BC089597 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145424.2 | NP_663399.2 | 4 | 846 | ||
GenBank mRNA | AI099357.1 | - | 2 | 227 | ||
AK144071.1 | - | 4 | 835 | |||
AY053571.1 | - | 4 | 766 | |||
BC018263.1 | - | 4 | 842 | 4239862 | ||
BC089597.1 | - | 4 | 819 | 30120654 |
