siRNA Details
| Gene Symbol: Ccm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001190343.1 | NP_001177272.1 | 9 | 1257 | ||
| NM_001190344.1 | NP_001177273.1 | 10 | 1343 | |||
| NM_146014.3 | NP_666126.1 | 10 | 1431 | |||
| GenBank mRNA | AK155145.1 | - | 10 | 1405 | ||
| AK166131.1 | - | 7 | 958 | |||
| AK169727.1 | - | 10 | 1345 | |||
| AK188818.1 | - | 2 | 256 | |||
| AY442689.1 | - | 10 | 1318 | |||
| BC029157.1 | - | 10 | 1362 | 4952288 | ||

Ccm2
Genomic Map