siRNA Details
| Gene Symbol: Clint1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001045520.3 | NP_001038985.2 | 10 | 1502 | ||
| GenBank mRNA | AB093212.1 | - | 8 | 2514 | ||
| AK143544.1 | - | 10 | 1503 | |||
| AK147875.1 | - | 10 | 1487 | |||
| AK167454.1 | - | 2 | 114 | |||
| AK168991.1 | - | 10 | 1468 | |||
| BC004080.1 | - | 7 | 1040 | |||

Clint1
Genomic Map