siRNA Details
| Gene Symbol: Clint1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001045520.3 | NP_001038985.2 | 11 | 1581 | ||
| GenBank mRNA | AB093212.1 | - | 8 | 4019 | ||
| AK143544.1 | - | 11 | 1582 | |||
| AK147875.1 | - | 11 | 1566 | |||
| AK167454.1 | - | 3 | 193 | |||
| AK168991.1 | - | 11 | 1547 | |||
| BC004080.1 | - | 8 | 1119 | |||

Clint1
Genomic Map