siRNA Details
| Gene Symbol: Wnt9a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139298.2 | NP_647459.1 | 4 | 677 | ||
| GenBank mRNA | AB072311.1 | - | 4 | 641 | ||
| AK087490.1 | - | 4 | 683 | |||
| BC066165.1 | - | 4 | 654 | 30435371 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Wnt9a| Gene Symbol: Wnt9a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139298.2 | NP_647459.1 | 4 | 677 | ||
| GenBank mRNA | AB072311.1 | - | 4 | 641 | ||
| AK087490.1 | - | 4 | 683 | |||
| BC066165.1 | - | 4 | 654 | 30435371 | ||