siRNA Details
| Gene Symbol: Nlrp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145827.3 | NP_665826.1 | 4 | 1723 | ||
| GenBank mRNA | AF486632.1 | - | 5 | 1619 | ||
| AY337285.1 | - | 3 | 1509 | |||
| AY355340.1 | - | 3 | 1497 | |||
| AY495376.1 | - | 4 | 1723 | |||
| AY495377.1 | - | 5 | 1619 | |||
| BC116174.1 | - | 4 | 1683 | 40048724 | ||
| BC116175.1 | - | 4 | 1683 | 40048725 | ||
| KF032621.1 | - | 3 | 1499 | |||

Nlrp3
Genomic Map