siRNA Details
| Gene Symbol: Trp53i13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024920.1 | NP_001020091.1 | 5 | 388 | ||
| GenBank mRNA | BC147324.1 | - | 5 | 386 | 9056369 | |
| BC147325.1 | - | 5 | 386 | 9056377 | ||
| BQ939386.1 | - | 5 | 388 | |||

Trp53i13
Genomic Map