siRNA Details
| Gene Symbol: Gm11545 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105561.1 | NP_001099031.1 | 1, 2 | 346 | ||
| GenBank mRNA | AK139527.1 | - | 1, 2 | 346 | ||
| AK169863.1 | - | 1, 2 | 119 | |||
| AK170015.1 | - | 1, 2 | 173 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm11545| Gene Symbol: Gm11545 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105561.1 | NP_001099031.1 | 1, 2 | 346 | ||
| GenBank mRNA | AK139527.1 | - | 1, 2 | 346 | ||
| AK169863.1 | - | 1, 2 | 119 | |||
| AK170015.1 | - | 1, 2 | 173 | |||