siRNA Details
| Gene Symbol: Cdc27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001285988.1 | NP_001272917.1 | 14 | 1999 | ||
| NM_001285989.1 | NP_001272918.1 | 14 | 1981 | |||
| NM_001285990.1 | NP_001272919.1 | 13 | 1904 | |||
| NM_145436.2 | NP_663411.2 | 14 | 1984 | |||
| GenBank mRNA | AK148278.1 | - | 14 | 1839 | ||
| AK161143.1 | - | 3 | 360 | |||
| AK161471.1 | - | 14 | 1850 | |||
| BC157955.1 | - | 14 | 1998 | 9088128 | ||
| BC172100.1 | - | 14 | 1998 | 9054386 | ||
| BC172149.1 | - | 13 | 1903 | |||

Cdc27
Genomic Map