siRNA Details
| Gene Symbol: Prss40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009356.2 | NP_033382.2 | 3 | 393 | ||
| GenBank mRNA | AB008911.1 | - | 3 | 376 | ||
| AK132909.1 | - | 3 | 423 | |||
| BC145989.1 | - | 3 | 408 | 40131292 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prss40| Gene Symbol: Prss40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009356.2 | NP_033382.2 | 3 | 393 | ||
| GenBank mRNA | AB008911.1 | - | 3 | 376 | ||
| AK132909.1 | - | 3 | 423 | |||
| BC145989.1 | - | 3 | 408 | 40131292 | ||