siRNA Details
| Gene Symbol: Susd6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001242419.1 | NP_001229348.1 | 4 | 744 | ||
| NM_178682.4 | NP_848797.1 | 3 | 681 | |||
| GenBank mRNA | AK030233.1 | - | 4 | 767 | ||
| AK054203.1 | - | 2 | 396 | |||
| AK129100.1 | - | 4 | 507 | |||
| AK143867.1 | - | 4 | 724 | |||
| AK151939.1 | - | 3 | 648 | |||
| AK152820.1 | - | 3 | 648 | |||
| AK161132.1 | - | 3 | 683 | |||
| AK164823.1 | - | 3 | 648 | |||
| BC040401.1 | - | 3 | 654 | 4193723 | ||

Susd6
Genomic Map