siRNA Details
| Gene Symbol: Ppp2r5d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009358.3 | NP_033384.2 | 6 | 808 | ||
| GenBank mRNA | AB053464.1 | - | 6 | 753 | ||
| AF316616.2 | - | 6 | 665 | |||
| AK154050.1 | - | 6 | 784 | |||
| AK160587.1 | - | 6 | 749 | |||
| AY338234.1 | - | 6 | 738 | |||
| BC010716.1 | - | 6 | 721 | 2655861 | ||

Ppp2r5d
Genomic Map