siRNA Details
| Gene Symbol: Cirh1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011574.2 | NP_035704.2 | 12 | 1452 | ||
| GenBank mRNA | AK144455.1 | - | 12 | 1437 | ||
| AK146263.1 | - | 12 | 1428 | |||
| AK150862.1 | - | 12 | 1428 | |||
| AK168470.1 | - | 12 | 1402 | |||
| AK173318.1 | - | 11 | 1393 | |||
| AK177750.1 | - | 2 | 210 | |||
| BC027399.1 | - | 8 | 951 | 4948207 | ||
| BC068113.1 | - | 12 | 1441 | 5707865 | ||

Cirh1a
Genomic Map