siRNA Details
| Gene Symbol: Nrde2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290303.1 | NP_001277232.1 | 3 | 265 | ||
| NM_183155.3 | NP_898978.2 | 3 | 250 | |||
| GenBank mRNA | BC051175.1 | - | 2 | 155 | ||
| CD546085.1 | - | 3 | 232 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nrde2| Gene Symbol: Nrde2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290303.1 | NP_001277232.1 | 3 | 265 | ||
| NM_183155.3 | NP_898978.2 | 3 | 250 | |||
| GenBank mRNA | BC051175.1 | - | 2 | 155 | ||
| CD546085.1 | - | 3 | 232 | |||