siRNA Details
| Gene Symbol: Nrde2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290303.1 | NP_001277232.1 | 11 | 2517 | ||
| NM_183155.3 | NP_898978.2 | 11 | 2502 | |||
| GenBank mRNA | BC002230.1 | - | 1 | 211 | ||
| BC025577.1 | - | 6 | 1122 | 5323598 | ||
| BC051175.1 | - | 10 | 2407 | |||
| CN710548.1 | - | 1 | 32 | |||
| DN174402.1 | - | 2 | 512 | |||

Nrde2
Genomic Map