siRNA Details
| Gene Symbol: Nrde2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290303.1 | NP_001277232.1 | 11 | 3075 | ||
| NM_183155.3 | NP_898978.2 | 11 | 3060 | |||
| GenBank mRNA | BC002230.1 | - | 1 | 769 | ||
| BC025577.1 | - | 6 | 1680 | 5323598 | ||
| BC051175.1 | - | 10 | 2965 | |||
| BG067060.2 | - | 4 | 644 | |||

Nrde2
Genomic Map