siRNA Details
| Gene Symbol: Rin3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001161365.1 | NP_001154837.1 | 6 | 1428 | ||
| NM_177620.4 | NP_808288.2 | 6 | 1539 | |||
| GenBank mRNA | AK032205.1 | - | 6 | 1540 | ||
| AK154506.1 | - | 6 | 1564 | |||
| AK154522.1 | - | 6 | 1343 | |||
| AK154963.1 | - | 6 | 1429 | |||
| AK166196.1 | - | 6 | 1385 | |||
| AK171301.1 | - | 6 | 1353 | |||
| AK171311.1 | - | 6 | 1344 | |||
| AY265356.1 | - | 6 | 1525 | |||
| BC137907.1 | - | 6 | 1467 | 8860926 | ||

Rin3
Genomic Map