siRNA Details
| Gene Symbol: Ifi27l2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145449.2 | NP_663424.1 | 4 | 282 | ||
| GenBank mRNA | BC021795.1 | - | 4 | 282 | 4217067 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ifi27l2b| Gene Symbol: Ifi27l2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145449.2 | NP_663424.1 | 4 | 282 | ||
| GenBank mRNA | BC021795.1 | - | 4 | 282 | 4217067 | |