siRNA Details
| Gene Symbol: Cep170b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024602.3 | NP_001019773.2 | 13 | 3961 | ||
| GenBank mRNA | AK122236.1 | - | 14 | 3890 | ||
| AK147410.1 | - | 13 | 3902 | |||
| AK147612.1 | - | 11 | 3566 | |||
| BC050077.1 | - | 10 | 1047 | |||

Cep170b
Genomic Map