siRNA Details
| Gene Symbol: Thrsp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009381.2 | NP_033407.1 | 1 | 427 | ||
| GenBank mRNA | AK050300.1 | - | 1 | 437 | ||
| AK077300.1 | - | 1 | 220 | |||
| BC009165.1 | - | 1 | 427 | 2811774 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Thrsp| Gene Symbol: Thrsp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009381.2 | NP_033407.1 | 1 | 427 | ||
| GenBank mRNA | AK050300.1 | - | 1 | 437 | ||
| AK077300.1 | - | 1 | 220 | |||
| BC009165.1 | - | 1 | 427 | 2811774 | ||