siRNA Details
| Gene Symbol: Rasa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145452.3 | NP_663427.2 | 17 | 2811 | ||
| GenBank mRNA | AK133354.1 | - | 13 | 1371 | ||
| AK161497.1 | - | 7 | 608 | |||
| BC013637.1 | - | 15 | 1557 | 3498774 | ||
| BC065082.1 | - | 17 | 2353 | |||

Rasa1
Genomic Map