siRNA Details
| Gene Symbol: Ankrd34b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175455.4 | NP_780664.2 | 5 | 744 | ||
| GenBank mRNA | AK078203.1 | - | 5 | 818 | ||
| AK090387.1 | - | 5 | 746 | |||
| AK138465.1 | - | 4 | 930 | |||
| BC096609.1 | - | 5 | 620 | |||
| BC119312.1 | - | 5 | 606 | 8734061 | ||
| BC119314.1 | - | 5 | 606 | 8734063 | ||

Ankrd34b
Genomic Map