siRNA Details
| Gene Symbol: Serinc5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172588.2 | NP_766176.1 | 2 | 239 | ||
| GenBank mRNA | AB029501.1 | - | 2 | 128 | ||
| AK037699.1 | - | 2 | 239 | |||
| AK054475.1 | - | 2 | 250 | |||
| AK136799.1 | - | 2 | 287 | |||
| AK163675.1 | - | 2 | 204 | |||
| AK165334.1 | - | 2 | 246 | |||
| BC049189.1 | - | 2 | 239 | 6311617 | ||
| BC062131.1 | - | 2 | 239 | 6514513 | ||

Serinc5
Genomic Map