siRNA Details
| Gene Symbol: Wdr41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172590.3 | NP_766178.2 | 13, 14 | 1419 | ||
| GenBank mRNA | AK046853.1 | - | 13, 14 | 1388 | ||
| AK149993.1 | - | 13, 14 | 1385 | |||
| AK183715.1 | - | 1, 2 | 50 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Wdr41| Gene Symbol: Wdr41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172590.3 | NP_766178.2 | 13, 14 | 1419 | ||
| GenBank mRNA | AK046853.1 | - | 13, 14 | 1388 | ||
| AK149993.1 | - | 13, 14 | 1385 | |||
| AK183715.1 | - | 1, 2 | 50 | |||