siRNA Details
| Gene Symbol: Ccno | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081062.1 | NP_001074531.1 | 1, 2 | 493 | ||
| GenBank mRNA | AK086507.1 | - | 1, 2 | 468 | ||
| BC147759.1 | - | 1, 2 | 469 | 9087484 | ||
| BC147766.1 | - | 1, 2 | 469 | 9087491 | ||

Ccno
Genomic Map