siRNA Details
| Gene Symbol: Fam208a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001114879.1 | NP_001108351.1 | 16 | 2850 | ||
| NM_028945.1 | NP_083221.1 | 16 | 2850 | |||
| GenBank mRNA | AK028620.1 | - | 3 | 601 | ||
| AK044563.1 | - | 16 | 2851 | |||
| BC037390.1 | - | 2 | 341 | |||

Fam208a
Genomic Map