siRNA Details
Gene Symbol: Tln1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011602.5 | NP_035732.2 | 55 | 7806 | ||
GenBank mRNA | AK122420.2 | - | 55 | 7432 | ||
AK147228.1 | - | 55 | 7377 | |||
AK170630.1 | - | 21 | 2849 | |||
AK171322.1 | - | 21 | 2849 | |||
BC018557.1 | - | 19 | 2514 | |||
BC121787.1 | - | 25 | 3318 | |||
BC150810.1 | - | 55 | 7477 | 9087721 | ||
X56123.1 | - | 55 | 7478 |
