siRNA Details
| Gene Symbol: Tlx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009392.2 | NP_033418.1 | 1 | 282 | ||
| GenBank mRNA | AK013072.1 | - | 1 | 282 | ||
| BC138240.1 | - | 1 | 156 | 8861259 | ||
| BC138241.1 | - | 1 | 156 | 8861260 | ||
| M75953.1 | - | 1 | 200 | |||

Tlx2
Genomic Map