siRNA Details
| Gene Symbol: Tnc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011607.3 | NP_035737.2 | 14 | 4437 | ||
| GenBank mRNA | D90343.1 | - | 14 | 4238 | ||
| X56304.1 | - | 13 | 4171 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tnc| Gene Symbol: Tnc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011607.3 | NP_035737.2 | 14 | 4437 | ||
| GenBank mRNA | D90343.1 | - | 14 | 4238 | ||
| X56304.1 | - | 13 | 4171 | |||