siRNA Details
| Gene Symbol: Pglyrp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009402.2 | NP_033428.1 | 1 | 165 | ||
| GenBank mRNA | AF076482.1 | - | 1 | 164 | ||
| AF193843.1 | - | 1 | 153 | |||
| AK008335.1 | - | 1 | 166 | |||
| AK043227.1 | - | 1 | 163 | |||
| BC005582.1 | - | 1 | 175 | 3969014 | ||
| CT010165.1 | - | 1 | 129 | |||
| X86374.1 | - | 1 | 164 | |||

Pglyrp1
Genomic Map