siRNA Details
| Gene Symbol: Tnks | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175091.3 | NP_780300.2 | 18 | 2705 | ||
| GenBank mRNA | AK048860.1 | - | 14 | 2070 | ||
| AK080913.1 | - | 18 | 2708 | |||
| BC057370.1 | - | 18 | 2686 | 6835784 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tnks| Gene Symbol: Tnks | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175091.3 | NP_780300.2 | 18 | 2705 | ||
| GenBank mRNA | AK048860.1 | - | 14 | 2070 | ||
| AK080913.1 | - | 18 | 2708 | |||
| BC057370.1 | - | 18 | 2686 | 6835784 | ||