siRNA Details
| Gene Symbol: Tns1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289895.1 | NP_001276824.1 | 19 | 3021 | ||
| NM_027884.3 | NP_082160.3 | 19 | 3019 | |||
| GenBank mRNA | CF171717.1 | - | 1 | 275 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tns1| Gene Symbol: Tns1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289895.1 | NP_001276824.1 | 19 | 3021 | ||
| NM_027884.3 | NP_082160.3 | 19 | 3019 | |||
| GenBank mRNA | CF171717.1 | - | 1 | 275 | ||