siRNA Details
| Gene Symbol: Tmem165 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011626.2 | NP_035756.2 | 5, 6 | 1172 | ||
| GenBank mRNA | AK205450.1 | - | 1 | 13 | ||
| BC037638.1 | - | 6, 7 | 953 | |||
| BC141081.1 | - | 5, 6 | 956 | 9055693 | ||
| M23568.1 | - | 5, 6 | 1103 | |||

Tmem165
Genomic Map