siRNA Details
| Gene Symbol: Tpmt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016785.2 | NP_058065.2 | 3 | 338 | ||
| GenBank mRNA | AF037043.1 | - | 3 | 247 | ||
| AF037044.1 | - | 2 | 198 | |||
| AF046887.1 | - | 3 | 324 | |||
| AK002335.1 | - | 3 | 245 | |||
| AK019205.1 | - | 3 | 243 | |||
| BC021598.1 | - | 2 | 225 | 5098857 | ||
| CF106558.1 | - | 3 | 260 | |||

Tpmt
Genomic Map